Eldon James Threaded NPT(M) to Hose Barb Fittings, Straight Adapter, Natural Kynar®
Threaded barb adapter design provides safe, difficult-to-break connection
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Single barb design tightly “grips” the tubing
  • Biopharma compatible and USP 661 and BPOG compliant
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other general-purpose applications. Quickly and easily transition from soft tubing to a system that accepts a threaded connection using threaded barb adapters. Threads provide a tight and secure connection; a single barb on the other end of the fitting creates a tight seal to keep the tubing in place. An anti-rotation device on the hose barb prevents tubing from rotating, minimizing kinks.
These natural Kynar® fittings provide excellent chemical, flame, gamma radiation, UV, and weather resistance. They also exhibit good mechanical properties, low moisture absorption, and good machinability.
2,700.00 - ₹9,300.00INR / Pkg of 10
30 variations of this product are available.
See All Product Options Below
30 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 2) (in) Fitting Material Thread Size NPT(M) (Port 1) (InchShort)
Availability Pricing
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Kynar®, 3/4" NPT(M) x 5/8" ID; 10/PK 5012369 Mfr # A12-10NK7
COMPARE
5/8 PVDF 3/4
₹7,600.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Kynar®, 3/4" NPT(M) x 3/4" ID; 10/PK 5012370 Mfr # A12-12NK7
COMPARE
3/4 PVDF 3/4
₹8,100.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Kynar®, 3/4" NPT(M) x 1" ID; 10/PK 5012371 Mfr # A12-16NK7
COMPARE
1 PVDF 3/4
₹8,600.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Kynar®, 3/4" NPT(M) x 1/4" ID; 10/PK 5012372 Mfr # A12-4NK7
COMPARE
1/4 PVDF 3/4
₹7,000.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Kynar®, 3/4" NPT(M) x 3/8" ID; 10/PK 5012373 Mfr # A12-6NK7
COMPARE
3/8 PVDF 3/4
₹7,200.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Kynar®, 3/4" NPT(M) x 1/2" ID; 10/PK 5012374 Mfr # A12-8NK7
COMPARE
1/2 PVDF 3/4
₹7,300.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Kynar®, 1" NPT(M) x 1" ID; 10/PK 5012375 Mfr # A16-16NK7
COMPARE
1 PVDF 1
₹9,300.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Kynar®, 1/8" NPT(M) x 5/32" ID; 10/PK 5012376 Mfr # A2-2.5NK7
COMPARE
5/32 PVDF 1/8
₹3,300.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Kynar®, 1/8" NPT(M) x 1/8" ID; 10/PK 5012377 Mfr # A2-2NK7
COMPARE
1/8 PVDF 1/8
₹2,700.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Kynar®, 1/8" NPT(M) x 3/16" ID; 10/PK 5012378 Mfr # A2-3NK7
COMPARE
3/16 PVDF 1/8
₹3,700.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Kynar®, 1/8" NPT(M) x 1/4" ID; 10/PK 5012379 Mfr # A2-4NK7
COMPARE
1/4 PVDF 1/8
₹3,700.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Kynar®, 1/8" NPT(M) x 5/16" ID; 10/PK 5012380 Mfr # A2-5NK7
COMPARE
5/16 PVDF 1/8
₹3,700.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Kynar®, 1/8" NPT(M) x 3/8" ID; 10/PK 5012381 Mfr # A2-6NK7
COMPARE
3/8 PVDF 1/8
₹4,300.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Kynar®, 1/4" NPT(M) x 5/32" ID; 10/PK 5012382 Mfr # A4-2.5NK7
COMPARE
5/32 PVDF 1/4
₹4,600.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Kynar®, 1/4" NPT(M) x 1/8" ID; 10/PK 5012383 Mfr # A4-2NK7
COMPARE
1/8 PVDF 1/4
₹4,600.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Kynar®, 1/4" NPT(M) x 3/16" ID; 10/PK 5012384 Mfr # A4-3NK7
COMPARE
3/16 PVDF 1/4
₹4,600.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Kynar®, 1/4" NPT(M) x 1/4" ID; 10/PK 5012385 Mfr # A4-4NK7
COMPARE
1/4 PVDF 1/4
₹4,600.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Kynar®, 1/4" NPT(M) x 5/16" ID; 10/PK 5012386 Mfr # A4-5NK7
COMPARE
5/16 PVDF 1/4
₹5,000.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Kynar®, 1/4" NPT(M) x 3/8" ID; 10/PK 5012387 Mfr # A4-6NK7
COMPARE
3/8 PVDF 1/4
₹4,800.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Kynar®, 1/4" NPT(M) x 1/2" ID; 10/PK 5012388 Mfr # A4-8NK7
COMPARE
1/2 PVDF 1/4
₹5,100.00INR / Pkg of 10
Showing 1 - 20 of 30