Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Kynar®, 1/8" NPT(M) x 3/8" ID; 10/PK
Representative image only.
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Kynar®, 1/8" NPT(M) x 3/8" ID; 10/PK
Eldon James | Mfr: A2-6NK7| Item: 5012381
Threaded barb adapter design provides safe, difficult-to-break connection
₹4,300.00INR / Pkg of 10
₹4,300.00INR / Pkg of 10
Buy It Together
Current Item
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Kynar®, 1/8" NPT(M) x 3/8" ID; 10/PK
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Kynar®, 1/8" NPT(M) x 3/8" ID; 10/PK
5012381
₹4,300.00INR / Pkg of 10
+ + + + =
Total Price 15800.00 ADD 5 ITEMS TO CART
Product Description
Key Features
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Single barb design tightly “grips” the tubing
  • Biopharma compatible and USP 661 and BPOG compliant
  • 1/8" NPT(M) x 3/8" hose barb
More About this Item
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other general-purpose applications. Quickly and easily transition from soft tubing to a system that accepts a threaded connection using threaded barb adapters. Threads provide a tight and secure connection; a single barb on the other end of the fitting creates a tight seal to keep the tubing in place. An anti-rotation device on the hose barb prevents tubing from rotating, minimizing kinks.
These natural Kynar® fittings provide excellent chemical, flame, gamma radiation, UV, and weather resistance. They also exhibit good mechanical properties, low moisture absorption, and good machinability.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Fitting MaterialPVDF
  • Connection TypeThreaded NPT(M) x Hose Barb
  • Number Of Ports2
  • Thread Size NPT(M) (Port 1) (InchShort)1/8
  • Barb Size / Tubing Id (Port 2) (in)3/8
  • Flow PatternStraight
  • ColorNatural
  • Min Temperature (° F)-31
  • Max Temperature (° F)275
  • Min Temperature (° C)-35
  • Max Temperature (° C)135
  • Regulatory ApprovalsNon-DEHP;Non-Phthalate;USP
  • DescriptionThreaded to Hose Barb Fitting, Straight Adapter, Natural Kynar®, 1/8" NPT(M) x 3/8" ID; 10/PK