Eldon James Threaded NPT(M) to Hose Barb Fittings, Straight Adapter, Animal-Derivative-Free Polypropylene
Threaded barb adapter design provides safe, difficult-to-break connection
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Single barb design tightly “grips” the tubing
  • Meets FDA requirements 21 CFR 177.1520(a)(3)(i) and (c)3.1a and RoHS compliant
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other general-purpose applications. Quickly and easily transition from soft tubing to a system that accepts a threaded connection using threaded barb adapters. Threads provide a tight and secure connection; a single barb on the other end of the fitting creates a tight seal to keep the tubing in place. An anti-rotation device on the hose barb prevents tubing from rotating, minimizing kinks.
These animal-derivative-free polypropylene fittings are radiation stable and resistant to solvents, chemicals, water, strong mineral acids, bases, and other inorganic environments. Not recommended for use with oxidizing agents.
1,400.00 - ₹2,500.00INR / Pkg of 10
30 variations of this product are available.
See All Product Options Below
30 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 2) (in) Fitting Material Thread Size NPT(M) (Port 1) (InchShort)
Availability Pricing
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Animal-Derivative-Free Polypropylene, 3/4" NPT(M) x 5/8 ID; 10/PK 5012429 Mfr # A12-10PP
COMPARE
5/8 Polypropylene 3/4
₹1,700.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Animal-Derivative-Free Polypropylene, 3/4" NPT(M) x 3/4" ID; 10/PK 5012430 Mfr # A12-12PP
COMPARE
3/4 Polypropylene 3/4
₹2,300.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Animal-Derivative-Free Polypropylene, 3/4" NPT(M) x 1" ID; 10/PK 5012431 Mfr # A12-16PP
COMPARE
1 Polypropylene 3/4
₹2,500.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Animal-Derivative-Free Polypropylene, 3/4" NPT(M) x 1/4" ID; 10/PK 5012432 Mfr # A12-4PP
COMPARE
1/4 Polypropylene 3/4
₹1,600.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Animal-Derivative-Free Polypropylene, 3/4" NPT(M) x 3/8" ID; 10/PK 5012433 Mfr # A12-6PP
COMPARE
3/8 Polypropylene 3/4
₹1,600.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Animal-Derivative-Free Polypropylene, 3/4" NPT(M) x 1/2" ID; 10/PK 5012434 Mfr # A12-8PP
COMPARE
1/2 Polypropylene 3/4
₹1,600.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Animal-Derivative-Free Polypropylene, 1" NPT(M) x 1" ID; 10/PK 5012435 Mfr # A16-16PP
COMPARE
1 Polypropylene 1
₹2,500.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Animal-Derivative-Free Polypropylene, 1/8" NPT(M) x 5/32" ID; 10/PK 5012436 Mfr # A2-2.5PP
COMPARE
5/32 Polypropylene 1/8
₹1,400.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Animal-Derivative-Free Polypropylene, 1/8" NPT(M) x 1/8" ID; 10/PK 5012437 Mfr # A2-2PP
COMPARE
1/8 Polypropylene 1/8
₹1,600.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Animal-Derivative-Free Polypropylene, 1/8" NPT(M) x 3/16" ID; 10/PK 5012438 Mfr # A2-3PP
COMPARE
1/8 Polypropylene 1/8
₹1,400.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Animal-Derivative-Free Polypropylene, 1/8" NPT(M) x 1/4" ID; 10/PK 5012439 Mfr # A2-4PP
COMPARE
1/4 Polypropylene 1/8
₹1,800.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Animal-Derivative-Free Polypropylene, 1/8" NPT(M) x 5/16" ID; 10/PK 5012440 Mfr # A2-5PP
COMPARE
5/16 Polypropylene 1/8
₹1,400.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Animal-Derivative-Free Polypropylene, 1/8" NPT(M) x 3/8" ID; 10/PK 5012441 Mfr # A2-6PP
COMPARE
3/8 Polypropylene 1/8
₹1,600.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Animal-Derivative-Free Polypropylene, 1/4" NPT(M) x 5/32" ID; 10/PK 5012442 Mfr # A4-2.5PP
COMPARE
5/32 Polypropylene 1/4
₹1,400.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Animal-Derivative-Free Polypropylene, 1/4" NPT(M) x 1/8" ID; 10/PK 5012443 Mfr # A4-2PP
COMPARE
1/8 Polypropylene 1/4
₹1,400.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Animal-Derivative-Free Polypropylene, 1/4" NPT(M) x 3/16" ID; 10/PK 5012444 Mfr # A4-3PP
COMPARE
3/16 Polypropylene 1/4
₹1,400.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Animal-Derivative-Free Polypropylene, 1/4" NPT(M) x 1/4" ID; 10/PK 5012445 Mfr # A4-4PP
COMPARE
1/4 Polypropylene 1/4
₹1,400.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Animal-Derivative-Free Polypropylene, 1/4" NPT(M) x 5/16" ID; 10/PK 5012446 Mfr # A4-5PP
COMPARE
5/16 Polypropylene 1/4
₹1,500.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Animal-Derivative-Free Polypropylene, 1/4" NPT(M) x 3/8" ID; 10/PK 5012447 Mfr # A4-6PP
COMPARE
3/8 Polypropylene 1/4
₹1,500.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Animal-Derivative-Free Polypropylene, 1/4" NPT(M) x 1/2" ID; 10/PK 5012448 Mfr # A4-8PP
COMPARE
1/2 Polypropylene 1/4
₹1,800.00INR / Pkg of 10
Showing 1 - 20 of 30