Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Animal-Derivative-Free Polypropylene, 1/4" NPT(M) x 5/32" ID; 10/PK
Representative image only.
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Animal-Derivative-Free Polypropylene, 1/4" NPT(M) x 5/32" ID; 10/PK
Eldon James | Mfr: A4-2.5PP| Item: 5012442
Threaded barb adapter design provides safe, difficult-to-break connection
₹1,400.00INR / Pkg of 10
₹1,400.00INR / Pkg of 10
Buy It Together
Current Item
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Animal-Derivative-Free Polypropylene, 1/4" NPT(M) x 5/32" ID; 10/PK
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Animal-Derivative-Free Polypropylene, 1/4" NPT(M) x 5/32" ID; 10/PK
5012442
₹1,400.00INR / Pkg of 10
+ + +
Cole-Parmer PVC Lab Tubing, 1/8" ID x 3/16" OD; 50 Ft
5012115
₹7,500.00INR / Pkg of 1
+ + =
Total Price 92100.00 ADD 6 ITEMS TO CART
Product Description
Key Features
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Single barb design tightly “grips” the tubing
  • Meets FDA requirements 21 CFR 177.1520(a)(3)(i) and (c)3.1a and RoHS compliant
  • 1/4" NPT(M) x 5/32" hose barb
More About this Item
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other general-purpose applications. Quickly and easily transition from soft tubing to a system that accepts a threaded connection using threaded barb adapters. Threads provide a tight and secure connection; a single barb on the other end of the fitting creates a tight seal to keep the tubing in place. An anti-rotation device on the hose barb prevents tubing from rotating, minimizing kinks.
These animal-derivative-free polypropylene fittings are radiation stable and resistant to solvents, chemicals, water, strong mineral acids, bases, and other inorganic environments. Not recommended for use with oxidizing agents.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Fitting MaterialPolypropylene
  • Number Of Ports2
  • Thread Size NPT(M) (Port 1) (InchShort)1/4
  • Barb Size / Tubing Id (Port 2) (in)5/32
  • Flow PatternStraight
  • ColorNatural
  • Min Temperature (° F)-9
  • Max Temperature (° F)150
  • Min Temperature (° C)-23
  • Max Temperature (° C)66
  • Regulatory ApprovalsFDA;RoHS;ADCF;Non-DEHP;Non-Phthalate
  • DescriptionThreaded to Hose Barb Fitting, Straight Adapter, Animal-Derivative-Free Polypropylene, 1/4" NPT(M) x 5/32" ID; 10/PK