Eldon James Hose Barb Fittings, Y Union, Stainless Steel
Excellent for high temperature and harsh physical environments
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Quick and easy assembly without tools
  • Single barb design tightly “grips” the tubing
  • RoHS compliant
Easily combine or split fluid flow using Y unions. These durable stainless steel fittings perform well in high temperatures and challenging environments. Made of 316L molybdenum bearing stainless steel, they are widely used in pharmaceutical, food, and beverage applications. A single barb on each end of the fitting creates a tight seal to keep the tubing in place.
3,400.00 - ₹5,100.00INR / Pkg of 10
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Barb Size / Tubing Id (Port 3) (in)
Availability Pricing
Eldon James Hose Barb Fitting, Y Union, Natural Kynar®, 3/32" ID; 10/PK 5012892 Mfr # Y0-1.5NK7
COMPARE
3/32 3/32 3/32
₹3,400.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Y Union, Natural Kynar®, 5/8" ID; 10/PK 5012893 Mfr # Y0-10NK7
COMPARE
5/8 5/8 5/8
₹21,000.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Y Union, Natural Kynar®, 3/4" ID; 10/PK 5012894 Mfr # Y0-12NK7
COMPARE
3/4 3/4 3/4
₹26,000.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Y Union, Natural Kynar®, 1" ID; 10/PK 5012895 Mfr # Y0-16NK7
COMPARE
1 1 1
₹28,500.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Y Union, Natural Kynar®, 1/16" ID; 10/PK 5012896 Mfr # Y0-1NK7
COMPARE
1/16 1/16 1/16
₹3,400.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Y Union, Natural Kynar®, 5/32" ID; 10/PK 5012897 Mfr # Y0-2.5NK7
COMPARE
5/32 5/32 5/32
₹5,100.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Y Union, Natural Kynar®, 1/8" ID; 10/PK 5012898 Mfr # Y0-2NK7
COMPARE
1/8 1/8 1/8
₹3,400.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Y Union, Natural Kynar®, 3/16" ID; 10/PK 5012899 Mfr # Y0-3NK7
COMPARE
3/16 3/16 3/16
₹5,100.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Y Union, Natural Kynar®, 1/4" ID; 10/PK 5012900 Mfr # Y0-4NK7
COMPARE
1/4 1/4 1/4
₹6,000.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Y Union, Natural Kynar®, 5/16" ID; 10/PK 5012901 Mfr # Y0-5NK7
COMPARE
5/16 5/16 5/16
₹14,100.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Y Union, Natural Kynar®, 3/8" ID; 10/PK 5012902 Mfr # Y0-6NK7
COMPARE
3/8 3/8 3/8
₹14,200.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Y Union, Natural Kynar®, 1/2" ID; 10/PK 5012903 Mfr # Y0-8NK7
COMPARE
1/2 1/2 1/2
₹15,900.00INR / Pkg of 10
Showing 1- 12 of 12