Eldon James Hose Barb Fitting, Y Union, Natural Kynar®, 5/32" ID; 10/PK
Representative image only.
Eldon James Hose Barb Fitting, Y Union, Natural Kynar®, 5/32" ID; 10/PK
Eldon James | Mfr: Y0-2.5NK7| Item: 5012897
Single barb design tightly “grips” tubing
₹5,100.00INR / Pkg of 10
₹5,100.00INR / Pkg of 10
Product Description
Key Features
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Quick and easy assembly without tools
  • Single barb design tightly “grips” the tubing
  • RoHS compliant
  • 5/32" hose barb
More About this Item
Easily combine or split fluid flow using Y unions. These durable stainless steel fittings perform well in high temperatures and challenging environments. Made of 316L molybdenum bearing stainless steel, they are widely used in pharmaceutical, food, and beverage applications. A single barb on each end of the fitting creates a tight seal to keep the tubing in place.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Fitting MaterialPVDF;PVDF
  • Sterilization MethodE-beam/gamma irradiation, EtO, or autoclave up to 135°C;E-beam/gamma irradiation, EtO, or autoclave up to 135°C
  • Connection TypeHose Barb Y;Hose Barb Y
  • Number Of Ports3;3
  • Barb Size / Tubing Id (Port 1) (in)5/32;5/32
  • Barb Size / Tubing Id (Port 2) (in)5/32;5/32
  • Barb Size / Tubing Id (Port 3) (in)5/32;5/32
  • Flow PatternY;Y
  • ColorNatural;Natural
  • Min Temperature (° F)-31;-31
  • Max Temperature (° F)275;275
  • Min Temperature (° C)-35;-35
  • Max Temperature (° C)135;135
  • Regulatory ApprovalsNon-DEHP;USP;Non-DEHP;Non-Phthalate;USP;Non-Phthalate
  • DescriptionHose Barb Fitting, Y Union, Natural Kynar®, 5/32" ID; 10/PK;Hose Barb Fitting, Y Union, Natural Kynar®, 5/32" ID; 10/PK