Ekman Dredge
Quickly sample sediment from various bodies of water
  • Ideal for collecting samples of bottom fauna for productivity, biological, and geological studies
Sample soft, silty, vegetation-free river, stream, or lake bottoms. Messenger-activated dredges are constructed of stainless steel with stainless steel springs and cables. Feature flow-through design for low drag and good descent stability. Dredges have positive-acting lid and jaw-control mechanism. Includes a marine plywood carrying case. Order required messenger and descent cable separately.
200,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Material Sample Volume
Availability Pricing
Wildco Ekman Dredge with Case, 6" x 6" x 6" Sample Area 0547000 Mfr # 196-B10
COMPARE
Stainless Steel
₹200,000.00INR / Each