ECO Funnel® Solvent Safety Funnels
Cap chemical waste containers to prevent contamination or fire
  • Proprietary design helps ensure safe chemical waste disposal in laboratories
  • Lids close and latch easily to keep fumes from escaping waste containers
  • Maintain EPA/OSHA compliance and greatly reduce risk of lab mishaps
Made of high-density polyethylene (HDPE), funnels are compatible with most chemical solvents and help keep emissions to near zero. Ideal containment system for eliminating the danger of ignited vapors and spills in chemical, biotech, chromatography and HPLC environments. Choose between funnel cap adapters or complete systems. Systems with bottles include a secondary container with detachable base.
50,200.00 - ₹95,600.00INR / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Diameter (InchShort) Compatible With Description
Availability Pricing
ECO Funnel Solvent Safety Funnel with FS-70 (Rieke® 8TPI) Cap Adapter; 4" dia. 8140101 Mfr # EF-4-FS70
COMPARE
4 70 mm (Rieke FS-70) Solvent Safety Funnel with FS-70 (Rieke® 8TPI) Cap Adapter; 4" dia.
₹63,600.00INR / Each
ECO Funnel Solvent Safety Funnel with 38-430 mm Cap Adapter; 4" dia. 8140102 Mfr # EF-4-38
COMPARE
4 38-430 Solvent Safety Funnel with 38-430 mm Cap Adapter; 4" dia.
₹50,200.00INR / Each
ECO Funnel Solvent Safety Funnel with 2" NPT Fine Thread Cap Adapter; 8" dia. 8140103 Mfr # EF-4716-1P
COMPARE
8 2" NPT(M) Solvent Safety Funnel with 2" NPT Fine Thread Cap Adapter; 8" dia.
₹63,600.00INR / Each
ECO Funnel Solvent Safety Funnel with FS-70 (Rieke® 8TPI) Cap Adapter; 8" dia. 8140104 Mfr # EF-8-FS70
COMPARE
8 70 mm (Rieke FS-70) Solvent Safety Funnel with FS-70 (Rieke® 8TPI) Cap Adapter; 8" dia.
₹63,600.00INR / Each
ECO Funnel Solvent Safety Funnel with 70 mm Nalgene® Cap Adapter; 8" dia. 8140106 Mfr # EF-30020
COMPARE
8 70 mm Nalgene® Solvent Safety Funnel with 70 mm Nalgene® Cap Adapter; 8" dia.
₹63,600.00INR / Each
ECO Funnel Solvent Safety Funnel with 83B Nalgene® Cap Adapter; 8" dia. 8140107 Mfr # EF-8-83B
COMPARE
8 83B Nalgene® Solvent Safety Funnel with 83B Nalgene® Cap Adapter; 8" dia.
₹74,100.00INR / Each
ECO Funnel Solvent Safety Funnel with 38-430 mm Cap Adapter; 8" dia. 8140108 Mfr # EF-3004C
COMPARE
8 38-430 Solvent Safety Funnel with 38-430 mm Cap Adapter; 8" dia.
₹50,200.00INR / Each
ECO Funnel Solvent Safety Funnel for Justrite® Poly Disposal Cans; 8" 8140109 Mfr # EF-8-Justrite-B
COMPARE
8 3.65" Justrite® poly disposal cans Solvent Safety Funnel for Justrite® Poly Disposal Cans; 8"
₹81,000.00INR / Each
ECO Funnel Solvent Safety Funnel System with 4-L LDPE Bottle, 38-430 mm Cap Adapter, Detachable Base; 8" dia. 8140111 Mfr # EF-3004C-SYS
COMPARE
8 38-430 Solvent Safety Funnel System with 4-L LDPE Bottle, 38-430 mm Cap Adapter, Detachable Base; 8" dia.
₹79,900.00INR / Each
ECO Funnel Solvent Safety Funnel System with 4-L LDPE Bottle, 38-430 mm Cap Adapter, Detachable Base; 4" dia. 8140112 Mfr # EF-4-38-SYS
COMPARE
4 38-430 Solvent Safety Funnel System with 4-L LDPE Bottle, 38-430 mm Cap Adapter, Detachable Base; 4" dia.
₹81,000.00INR / Each
ECO Funnel Solvent Safety Funnel System with 5-gal. Drum, FS-70 (Rieke® 8TPI) Cap Adapter; 8" dia. 8140113 Mfr # EF-8-FS70-SYS
COMPARE
8 70 mm (Rieke FS-70) Solvent Safety Funnel System with 5-gal. Drum, FS-70 (Rieke® 8TPI) Cap Adapter; 8" dia.
₹95,600.00INR / Each
Showing 1- 11 of 11