ECO Funnel Solvent Safety Funnel with 2" NPT Fine Thread Cap Adapter; 8" dia.
Representative image only.
ECO Funnel Solvent Safety Funnel with 2" NPT Fine Thread Cap Adapter; 8" dia.
| Mfr: EF-4716-1P| Item: 8140103
Cap chemical waste containers to prevent contamination or fire
₹63,600.00INR / Each
₹63,600.00INR / Each
Buy It Together
Current Item
ECO Funnel Solvent Safety Funnel with 2" NPT Fine Thread Cap Adapter; 8" dia.
ECO Funnel Solvent Safety Funnel with 2" NPT Fine Thread Cap Adapter; 8" dia.
8140103
₹63,600.00INR / Each
+ =
Total Price 73500.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Proprietary design helps ensure safe chemical waste disposal in laboratories
  • Lids close and latch easily to keep fumes from escaping waste containers
  • Maintain EPA/OSHA compliance and greatly reduce risk of lab mishaps
  • Cap Adapter: 2" NPT Fine Thread Diameter of Funnel: 8"
More About this Item
Made of high-density polyethylene (HDPE), funnels are compatible with most chemical solvents and help keep emissions to near zero. Ideal containment system for eliminating the danger of ignited vapors and spills in chemical, biotech, chromatography and HPLC environments. Choose between funnel cap adapters or complete systems. Systems with bottles include a secondary container with detachable base.
Specifications
  • MaterialHDPE Funnel
  • Compatible With2" NPT(M)
  • Diameter (InchShort)8
  • DescriptionSolvent Safety Funnel with 2" NPT Fine Thread Cap Adapter; 8" dia.