EchoTherm Programmable Digital Stirring Hot Plates
Store up to 10 programs in memory for instant recall and use
  • Hot plate heats up to 450°C in approximately 2 minutes
  • Platinum RTD temperature measurement for ±1% accuracy
Repeat any program up to 98 times automatically. Multiple temperatures, temperature ramp rates, stirring speeds and timed events can be stored. Includes timer to 99 hours, 59 minutes with auto-off option. Unit can be programmable or non-programmable in operation. Offers RS232 interface for control or recording data via a computer. 6-foot cord with plug is included.
379,000.00 - ₹541,000.00INR / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Top Plate Length (in) Top Plate Material
Availability Pricing
Torrey Pines EchoTherm HS40 Programmable Digital Stirring Hot Plate, 115V 0464600 Mfr # HS40
COMPARE
120 8 Ceramic
₹474,000.00INR / Each
Torrey Pines EchoTherm HS40-2 Programmable Digital Stirring Hot Plate, 230V 0464610 Mfr # HS40-2
COMPARE
230 8 Ceramic
₹474,000.00INR / Each
Torrey Pines EchoTherm HS60 Stirring Hot Plate, Programmable Ceramic, 6" x 6", 115V 5180871 Mfr # HS60
COMPARE
115 6 Ceramic
₹379,000.00INR / Each
Torrey Pines EchoTherm HS60-2 Stirring Hot Plate, Programmable Ceramic, 6" x 6", 230V 5180873 Mfr # HS60-2
COMPARE
230 6 Ceramic
₹379,000.00INR / Each
Torrey Pines EchoTherm HS60 Stirring Hot Plate, Programmable Aluminum, 6" x 6", 115V 5180875 Mfr # HS60A
COMPARE
115 6 Aluminum
₹391,000.00INR / Each
Torrey Pines EchoTherm HS60-2 Stirring Hot Plate, Programmable Aluminum, 6" x 6", 230V 5180877 Mfr # HS60A-2
COMPARE
230 6 Aluminum
₹391,000.00INR / Each
Torrey Pines EchoTherm HS40 Stirring Hot Plate, Programmable Ceramic, 12" x 12", 115V 5180879 Mfr # HS61
COMPARE
120 12 Ceramic
₹527,000.00INR / Each
Torrey Pines EchoTherm HS40 Stirring Hot Plate, Programmable Ceramic, 12" x 12", 230V 5180881 Mfr # HS61-2
COMPARE
230 12 Ceramic
₹527,000.00INR / Each
Torrey Pines EchoTherm HS40 Stirring Hot Plate, Programmable Aluminum, 12" x 12", 115V 5180883 Mfr # HS61A
COMPARE
120 12 Aluminum
₹541,000.00INR / Each
Torrey Pines EchoTherm HS40 Stirring Hot Plate, Programmable Aluminum, 12" x 12", 230V 5180885 Mfr # HS61A-2
COMPARE
230 12 Aluminum
₹541,000.00INR / Each
Showing 1- 10 of 10