EchoTherm Programmable Digital Hot Plates
Store up to 10 programs in memory for instant recall and use
  • Quick heat-up time — maximum temperature is reached in approximately 2 minutes
  • Precision platinum RTD temperature sensor for ±1% accuracy
Fully programmable hot plate allows you to program and store multiple temperatures, ramp rates, and timed events. Once programmed, routines can be recalled and repeated from 1 to 98 times or infinitely if desired. When a program is running, the unit will automatically monitor to ensure it runs exactly as programmed—offering totally hands-free and worry-free operation so you can focus on other tasks!
Features timed operation to 99 hours, 59 minutes with auto-off option. Unit can be programmable or non-programmable in operation. Offers RS-232 interface for control or recording data via a computer. Choose between a chemical-resistant ceramic top plate or milled aluminum for fast and accurate heat disbursement.
335,000.00 - ₹513,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Top Plate Length (in)
Availability Pricing
Torrey Pines EchoTherm HP60 Hot Plate, Programmable Ceramic, 6" x 6", 115V 5180891 Mfr # HP60
COMPARE
115 6
₹335,000.00INR / Each
Torrey Pines EchoTherm HP60-2 Hot Plate, Programmable Ceramic, 6" x 6", 230V 5180893 Mfr # HP60-2
COMPARE
230 6
₹335,000.00INR / Each
Torrey Pines EchoTherm HP61 Hot Plate, Programmable Ceramic, 12" x 12", 115V 5180899 Mfr # HP61
COMPARE
115 12
₹513,000.00INR / Each
Torrey Pines EchoTherm HP61-2 Hot Plate, Programmable Ceramic, 12" x 12", 230V 5180901 Mfr # HP61-2
COMPARE
230 12
₹513,000.00INR / Each