EAR Yellow Neon Earplugs
E.A.R. Yellow Neon Metal Detectable: Made of soft foam to provide the highest NRR available. Bright yellow neon color makes monitoring use easy. Metal detectable.
Compliance: ANSI S3.19-1974
13,400.00INR / Box of 200
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
NRR (dB)
Availability Pricing
3M E-A-R Yellow Neon Earplugs, Regular/uncorded 4849160 Mfr # 312-1250 (MODEL # NEON)
COMPARE
33
₹13,400.00INR / Box of 200