3M E-A-R Yellow Neon Earplugs, Regular/uncorded
Representative image only.
3M E-A-R Yellow Neon Earplugs, Regular/uncorded
3M | Mfr: 312-1250 (MODEL # NEON)| Item: 4849160
₹13,400.00INR / Box of 200
₹13,400.00INR / Box of 200
Product Description
More About this Item
E.A.R. Yellow Neon Metal Detectable: Made of soft foam to provide the highest NRR available. Bright yellow neon color makes monitoring use easy. Metal detectable.
Compliance: ANSI S3.19-1974
Specifications
  • NRR (dB)33
  • ComplianceANSI S3.19-1974
  • CordedNo
  • DescriptionEarplugs, Regular/uncorded