Eagle Space-Saving Flammable Liquid Safety Cabinets
Ideal where storage space is limited
  • Trilingual warning label
  • FDA approved
  • NFPA Code 30 and OSHA compliant
Slim-designed safety cabinets let you maximize storage space of flammables liquids and holds up to 24 gallons (91 L). They are constructed of double-walled, 18-gauge steel with 1 1/2" (3.8 cm) insulating air barrier, and internal and external welding. Includes two vents, each with 2” (5.1 cm) threaded fittings and a fire baffle and cap. Cabinets have a high gloss powder coat finish with red trilingual warning, plus grounding attachment, 3-point latch system, and 2-in (5.1 cm) raised, leakproof door sill. Each shelf is adjustable on 3 in (7.6 cm) centers, supported by 4 brackets, with a 350 lbs (159 kg) safe allowable load. Warning on door reads FLAMMABLE—KEEP FIRE AWAY.
199,000.00 - ₹322,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Capacity (Gal) Number of Shelves Self-Closing
Availability Pricing
Eagle 1923X Space-Saver Flammable Liquid Cabinet, Manual Close, 1 Door, 2 Shelves, 24 Gal WW-86393-00 Mfr # 1923X
COMPARE
24 3 No
₹322,000.00INR / Each
Eagle 2310X Space-Saver Flammable Liquid Cabinet, Self Close, 1 Door, 3 Shelves, 24 Gal WW-86393-02 Mfr # 2310X
COMPARE
24 3 Yes
₹199,000.00INR / Each