Eagle Drum Funnel and Cover

Reduces splashing
  • EPA 40 CFR 265.173 compliant
Both the drum funnel and cover are constructed of yellow high density polyethylene (HDPE). The drum funnel offers 6" high sidewalls to contain spills and splashing. The 18" wide funnel fits all 30- and 55-gallon drums and spout fits into any 2" NPT bung.

The cover fits the Eagle drum funnel and helps to keep out dirt and contaminants.

Note: Funnel and funnel cover are sold separately.
5,900.00 - ₹14,900.00INR / Each

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Eagle 1660 HDPE Drum Funnel, Spout for 2" NPT Bungs, Yellow 8611030 Mfr # 1660
COMPARE
Yellow HDPE Drum Funnel, Spout for 2" Bungs
₹14,900.00INR / Each
Eagle 1664 Drum Funnel Cover, Yellow, 18.5" Diameter 8611032 Mfr # 1664
COMPARE
Yellow HDPE Drum Funnel Cover
₹5,900.00INR / Each
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat