DWK Life Sciences (Kimble) MARK-M® Disposable Borosilicate Glass Tubes with Vertical Label
Write on 1-3/8"H marking area with marker or pencil for easy sample identification
  • USP Type I, ASTM E 438 Type I, Class B compliant
  • Manufactured from borosilicate glass 5.1
  • 1-3/8"H ceramic marking area is located 3/4" from the open end
  • Sturdy, uniform bottoms and consistent lengths
These disposable tubes are manufactured from borosilicate glass 5.1 to provide excellent chemical resistance. Easily label your samples for quick identification on a large, permanent ceramic marking area.
25,800.00 - ₹45,600.00INR / Case of 1000
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Special Features Tube Capacity (mL) Tube Outside Diameter (OD) (mm)
Availability Pricing
DWK Life Sciences (Kimble) MARK-M® Borosilicate Glass Tubes, Blue Marking Label, 10 x 75 mm, 3 mL; 1000/CS WW-34595-44 Mfr # 60A10BZB
COMPARE
Blue marking label, 1-3/8\"H 3 10
₹26,000.00INR / Case of 1000
DWK Life Sciences (Kimble) MARK-M® Borosilicate Glass Tubes, Green Marking Label, 10 x 75 mm, 3 mL; 1000/CS WW-34595-45 Mfr # 60A10BZG
COMPARE
Green marking label, 1-3/8\"H 3 10
₹29,600.00INR / Case of 1000
DWK Life Sciences (Kimble) MARK-M® Borosilicate Glass Tubes, Yellow Marking Label, 10 x 75 mm, 3 mL; 1000/CS WW-34595-46 Mfr # 60A10BZL
COMPARE
Yellow marking label, 1-3/8\"H 3 10
₹25,800.00INR / Case of 1000
DWK Life Sciences (Kimble) MARK-M® Borosilicate Glass Tubes, White Marking Label, 10 x 75 mm, 3 mL; 1000/CS WW-34595-47 Mfr # 60A10BZW
COMPARE
White marking label, 1-3/8\"H 3 10
₹25,800.00INR / Case of 1000
DWK Life Sciences (Kimble) MARK-M® Borosilicate Glass Tubes, Blue Marking Label, 12 x 75 mm, 5 mL; 1000/CS WW-34595-49 Mfr # 60B12BZB
COMPARE
Blue marking label, 1-3/8\"H 5 12
₹27,400.00INR / Case of 1000
DWK Life Sciences (Kimble) MARK-M® Borosilicate Glass Tubes, Green Marking Label, 12 x 75 mm, 5 mL; 1000/CS WW-34595-50 Mfr # 60B12BZG
COMPARE
Green marking label, 1-3/8\"H 5 12
₹27,400.00INR / Case of 1000
DWK Life Sciences (Kimble) MARK-M® Borosilicate Glass Tubes, Yellow Marking Label, 12 x 75 mm, 5 mL; 1000/CS WW-34595-51 Mfr # 60B12BZL
COMPARE
Yellow marking label, 1-3/8\"H 5 12
₹27,400.00INR / Case of 1000
DWK Life Sciences (Kimble) MARK-M® Borosilicate Glass Tubes, White Marking Label, 12 x 75 mm, 5 mL; 1000/CS WW-34595-52 Mfr # 60B12BZW
COMPARE
White marking label, 1-3/8\"H 5 12
₹27,400.00INR / Case of 1000
DWK Life Sciences (Kimble) MARK-M® Borosilicate Glass Tubes, White Marking Label, 16 x 100 mm, 14 mL; 1000/CS WW-34595-55 Mfr # 60E16BZW
COMPARE
White marking label, 1-3/8\"H 14 16
₹45,600.00INR / Case of 1000
Showing 1- 9 of 9