Drierite Relative Humidity Indicator Cards
Easy detects presence of moisture
  • Changes color when moisture content rises
  • Indicates relative humidity within desiccating chamber
  • Reuseable
These relative humidity cards easily indicate the presence of moisture in the air when their color changes from blue to pink. The cards can be reused. Just place the cards in a dry environment until they return to blue.
The cards are ideal for use in conjunction with the compartment desiccator, mini-dehumidifier, and desiccant bags. Place the cards in a desiccating chamber to indicate the relative humidity within the chamber. Two types of cards are available.
The 30% RH card changes color when the moisture content rises to 30% RH. The 10-60% RH card has 6 spots corresponding to 10, 20, 30, 40, 50, and 60% RH that change color individually when the moisture content reaches the indicated level.
180.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Drierite 60130 Relative Humidity Indicator Card at 30% RH 0893434 Mfr # 60130
COMPARE
Relative Humidity Indicator Card at 30% RH
₹180.00INR / Each
Drierite 60136 Relative Humidity Indicator Card, 6 Levels for 10% to 60% RH 0893435 Mfr # 60136
COMPARE
Relative Humidity Indicator Card, 6 Levels for 10% to 60% RH
₹180.00INR / Each