Diversey Soft Care Med Plus Hand Disinfectant
IPA based hand disinfectant effective against virus, bacteria, fungi, yeast & mold
  • Soft Care Med Plus is a ready-to-use, alcohol based hand spray
  • This is a leave-on product and does not need water or hand wash stations
  • 50% n-propanol and 20% IPA:
  • - Combinations are more potent than individual alcohol ingredient
  • Instant hand disinfection
  • Combination of 70% alcohol and 2.5% v/v CHG:
  • - Instant hand hygiene with residual sanitation - Wider microbial kill
Benefits:
Effective against vegetative bacteria, fungi, yeasts and viruses
Description:
Soft Care Med Plus Spray is an alcohol rub, developed for hand disinfection. The
product is suitable for application in the Pharma , healthcare and food processing area.
6,400.00 - ₹7,500.00INR / Case of 2
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Diversey Soft Care Med Plus Hand Spray 5 L; 2/Case 1639313 Mfr # 6284331
COMPARE
Diversey Soft Care Med Plus Hand Spray 5 L; 2/Case
₹6,400.00INR / Case of 2
Diversey Soft Care Med Plus Hand Spray, 500 mL; 20/Case 1639314 Mfr # 6284403
COMPARE
Diversey Soft Care Med Plus Hand Spray, 500 mL; 20/Case
₹7,500.00INR / Case of 20