Diversey Soft Care Des E Plus Hand Disinfectant
Ethanol based hand rub-in disinfectant effective against virus, bacteria, fungi, yeast & mold
  • Soft Care Des E plus is a ready-to-use, alcohol based hand rub.
  • This is a leave-on product and does not need water or hand wash stations
  • 70% Ethanol and 2.5% CHG:
  • - Instant hand hygiene with residual effect
  • - Wider microbial kill
  • Moisturiser and Emollients:
  • - Retain lipid and cellulose balance preventing Skin Irritation and Dehydration
  • - Soft, smooth and hygienic skin
Soft Care Des E Plus is an alcohol rub, developed for hand disinfection. The product is suitable for application in the healthcare and food processing area. Effective against virus, bacteria, fungi, yeast & mold.
5,500.00 - ₹6,600.00INR / Case of 20
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Diversey Softcare Des E Plus Hand Rub, 500 mL; 20/Case 1639311 Mfr # 6157076
COMPARE
Diversey Soft Care Des E Plus Hand Rub, 500 mL; 20/Case
₹6,600.00INR / Case of 20
Diversey Softcare Des E Plus Hand Rub, 5 L; 2/Case 1639312 Mfr # 6284340
COMPARE
Diversey Soft Care Des E Plus Hand Rub, 5 L; 2/Case
₹5,500.00INR / Case of 2