Distilling Apparatus for Kjeldahl Determinations
Distilling apparatus for Kjeldahl determinations is used for determining nitrogen in organic compounds. Perform both digestion and distillation steps in the flask. Includes 200-mm Graham condenser, 500-mL Kjeldahl flask, and connecting tube.
143,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Material Power (VAC) Description
Availability Pricing
Pyrex 3340-500 Distilling Apparatus for Kjeldahl Determinations 3453010 Mfr # 3340-500
COMPARE
Distilling Apparatus for Kjeldahl Determinations
₹143,000.00INR / Each