Pyrex 3340-500 Distilling Apparatus for Kjeldahl Determinations
Representative image only.
Pyrex 3340-500 Distilling Apparatus for Kjeldahl Determinations
Pyrex | Mfr: 3340-500| Item: 3453010
₹143,000.00INR / Each
₹143,000.00INR / Each
Product Description
More About this Item
Distilling apparatus for Kjeldahl determinations is used for determining nitrogen in organic compounds. Perform both digestion and distillation steps in the flask. Includes 200-mm Graham condenser, 500-mL Kjeldahl flask, and connecting tube.
Specifications
  • DescriptionDistilling Apparatus for Kjeldahl Determinations