Dilvac Stainless Steel Dewar Flasks
Safely store and transport low-temperature liquid gases
  • Specifically designed for applications where glass cannot be used
  • Safe to use with liquid nitrogen and liquid oxygen
These vacuum-insulted stainless steel Dewar flasks are designed for ultra-low or very high temperature applications. Constructed entirely of stainless steel, these flasks are rugged, light, and compact.
121,000.00 - ₹297,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Inside Diameter (ID) (in) Volume (Liters) Description
Availability Pricing
COMPARE
2 1/2 0.5 Stainless Steel Dewar, 500 mL
₹121,000.00INR / Each
COMPARE
3 19/64 1 Stainless Steel Dewar, 1 L
₹148,000.00INR / Each
COMPARE
4 1 Stainless Steel Dewar, 1 L
₹140,000.00INR / Each
COMPARE
4 2 Stainless Steel Dewar, 2 L
₹168,000.00INR / Each
COMPARE
7 19/64 3 Stainless Steel Dewar, 3 L
₹255,000.00INR / Each
COMPARE
7 19/64 6 Stainless Steel Dewar, 6 L
₹297,000.00INR / Each