Digi-Sense Thermocouple Mini Connector
Jacks and plugs good to 300°F (150°C)
  • Glass-filled nylon construction with hollow pins and molded barriers prevent short circuit
  • Secure thermocouple wire into positive/negative terminals with screws
  • Each connector comes in ANSI color coded material: type J-black, type K-yellow, and type T-blue
  • Miniature connectors are suited for small wires and mineral insulated cables
1,600.00 - ₹4,900.00INR / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Probe Type Description
Availability Pricing
COMPARE
300 K Miniature Type-K Thermocouple Male Connector, 2 Pin
₹1,600.00INR / Each
COMPARE
300 K Miniature Type-K Thermocouple Male Connector, 2 Pin, 5Pk
₹4,900.00INR / Pkg of 5
COMPARE
300 J Miniature Type-J Thermocouple Male Connector, 2 Pin
₹1,600.00INR / Each
COMPARE
300 J Miniature Type-J Thermocouple Male Connector, 2 Pin, 5Pk
₹4,900.00INR / Pkg of 5
COMPARE
300 T Miniature Type-T Thermocouple Male Connector, 2 Pin
₹1,600.00INR / Each
COMPARE
300 T Miniature Type-T Thermocouple Male Connector, 2 Pin, 5Pk
₹4,900.00INR / Pkg of 5
COMPARE
300 K Miniature Type-K Thermocouple Female Connector, 2 Pin
₹1,600.00INR / Each
COMPARE
300 K Miniature Type-K Thermocouple Female Connector, 2 Pin, 5Pk
₹4,900.00INR / Pkg of 5
COMPARE
300 J Miniature Type-J Thermocouple Female Connector, 2 Pin
₹1,600.00INR / Each
COMPARE
300 J Miniature Type-J Thermocouple Female Connector, 2 Pin, 5Pk
₹4,900.00INR / Pkg of 5
COMPARE
300 T Miniature Type-T Thermocouple Female Connector, 2 Pin
₹1,600.00INR / Each
Showing 1- 11 of 11