Digi-Sense Spade Lug Connectors
Secure thermocouple wire terminals
  • Maintain accuracy by choosing spade lugs that match the composition of your thermocouple wires
  • Available in the most common reference materials
  • Avoid confusion — each connector has the material abbreviation stamped into it
  • Sold in packs of ten
4,700.00 - ₹5,000.00INR / Pkg of 10
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Spade Lugs, Alumel, for Type K Thermocouples; 10/pk
₹5,000.00INR / Pkg of 10
COMPARE
Spade Lugs, Chromel, for Type K and E Thermocouples; 10/pk
₹4,900.00INR / Pkg of 10
COMPARE
Spade Lugs, Constantan, for Type J, T, and E Thermocouples; 10/pk
₹5,000.00INR / Pkg of 10
COMPARE
Spade Lugs, Copper, for Type T Thermocouples; 10/pk
₹4,700.00INR / Pkg of 10
COMPARE
Spade Lugs, Iron, for Type J Thermocouples; 10/pk
₹4,900.00INR / Pkg of 10