Digi-Sense Spade Lugs, Constantan, for Type J, T, and E Thermocouples; 10/pk
Representative image only.
Digi-Sense Spade Lugs, Constantan, for Type J, T, and E Thermocouples; 10/pk
Digi-Sense | Item: 1852804
Secure thermocouple wire terminals
₹5,000.00INR / Pkg of 10
₹5,000.00INR / Pkg of 10
Product Description
Key Features
  • Maintain accuracy by choosing spade lugs that match the composition of your thermocouple wires
  • Available in the most common reference materials
  • Avoid confusion — each connector has the material abbreviation stamped into it
  • Sold in packs of ten
  • 6 month warranty
6 Month Warranty
Specifications
  • DescriptionSpade Lugs, Constantan, for Type J, T, and E Thermocouples; 10/pk