Digi-Sense Red Spirit Oven Glass Thermometers, Celsius-scale
Replace mercury thermometers for a specific application with the same accuracy without the any hazards
  • Non-mercury liquid
These thermometers are designed for use in specific applications. These Durac thermometers feature markings that are permanently printed on the glass, individually serialized, and red spirit liquid that has a dark scale contrasts against a white background. Some thermometers are PTFE coated for added safety which may cause a slight change in accuracy up to 1 division.
Oven thermometers - designed for high temperature applications and feature a round glass swell to hold the thermometer in place
Call For Price
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Min Temperature (° C) Overall Length (mm)
Availability Pricing
COMPARE
130 20 135
Call For Price
COMPARE
130 20 135
Call For Price
COMPARE
200 35 180
Call For Price
COMPARE
200 35 180
Call For Price
COMPARE
200 0 280
Call For Price
COMPARE
200 0 280
Call For Price
COMPARE
200 0 400
Call For Price
COMPARE
200 0 400
Call For Price
Showing 1- 8 of 8