Digi-Sense Liquid-In-Glass Oven Thermometer; 35 to 200C, Organic Liquid Fill, Total Immersion

Representative image only.

Digi-Sense Liquid-In-Glass Oven Thermometer; 35 to 200C, Organic Liquid Fill, Total Immersion

Digi-Sense | Item: 0800647

Replace mercury thermometers for a specific application with the same accuracy without the any hazards

Call For Price

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

Call For Price

Buy It Together

Current Item

Digi-Sense Liquid-In-Glass Oven Thermometer; 35 to 200C, Organic Liquid Fill, Total Immersion
Digi-Sense Liquid-In-Glass Oven Thermometer; 35 to 200C, Organic Liquid Fill, Total Immersion
0800647

Call For Price
+ =
Total Price 33100.00 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Non-mercury liquid
  • 5 year warranty

More About this Item

These thermometers are designed for use in specific applications. These Durac thermometers feature markings that are permanently printed on the glass, individually serialized, and red spirit liquid that has a dark scale contrasts against a white background. Some thermometers are PTFE coated for added safety which may cause a slight change in accuracy up to 1 division.

Oven thermometers - designed for high temperature applications and feature a round glass swell to hold the thermometer in place

5 Year Warranty
Specifications
  • Length (")7 1/10
  • Min Temperature (° C)35
  • Max Temperature (° C)200
  • Accuracy±1°C, ±2°C above 100°C
  • Division (° C)1
  • Thermometer FillRed Spirit
  • Overall Length (InchShort)7.1
  • Overall Length (mm)180
  • Immersion Depth (mm)180
  • Stem MaterialWhite-Back Glass
  • DescriptionLiquid-In-Glass Oven Thermometer; 35 to 200C, Organic Liquid Fill, Total Immersion
  • Warranty5 years
Documents & Certificates
Reviews
Q&A