Digi-Sense Quick Disconnect Thermocouple Probe w/Standard Connectors - Ungrounded
Designed for everyday needs - probe comes installed with a quick disconnect connector and a mating female connector
  • Ideal for measuring liquids, gases, or semi solids
  • Designed with 316 Stanless Steel sheath
  • Response Time: 1.2 - 5 seconds or greater
  • Includes a stand alone female connector for instalation to a thermocouple extension cable
  • Each probe comes with a color coded ANSI dual standard connector: type J-black, type K-yellow, type T-blue, and type E-purple.
  • Custom designs or changes to this product are available upon request.
Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220°F (104°C); PVC handles are rated to 200°F (93°C); Stainless Steel handles are rated at 450°F (232°C); Mini Connectors are rated to 392°F (200°C)
7,200.00 - ₹11,200.00INR / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Probe Type Sheath Length (in)
Availability Pricing
COMPARE
1200 K 6
₹7,200.00INR / Each
COMPARE
1200 J 6
₹7,500.00INR / Each
COMPARE
1400 J 6
₹10,300.00INR / Each
COMPARE
500 T 6
₹10,300.00INR / Each
COMPARE
1200 J 12
₹7,700.00INR / Each
COMPARE
1400 J 12
₹10,700.00INR / Each
COMPARE
1400 J 12
₹11,200.00INR / Each
COMPARE
1200 J 18
₹7,900.00INR / Each
COMPARE
1650 K 18
₹8,700.00INR / Each
COMPARE
1650 K 18
₹9,200.00INR / Each
Showing 1- 10 of 10