Digi-Sense Panel-Mount Thermocouple Jacks
Round or rectangular shape to fit your instrument panel
  • Glass-filled nylon construction provides rigidity, strength, and surface hardness
  • Molded barriers prevent short circuit
Mount connectors to your panel using the two screws located on the front of the jack. Each connector is ANSI color-coded by thermocouple type: black (J), yellow (K), or blue (T). Choose between flat-bladed miniconnectors and round-pinned standard connectors.
990.00 - ₹1,700.00INR / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Panel-Mount Miniconnector, Type-J, Round, Female, 1/Ea
₹1,400.00INR / Each
COMPARE
Panel-Mount Miniconnector, Type-K, Round, Female, 1/Ea
₹1,400.00INR / Each
COMPARE
Panel-Mount Miniconnector, Type-T, Round, Female, 1/Ea
₹1,400.00INR / Each
COMPARE
Panel-Mount Standard Connector, Type-J, Round, Female, 1/Ea
₹1,700.00INR / Each
COMPARE
Panel-Mount Standard Connector, Type-K, Round, Female, 1/Ea
₹1,700.00INR / Each
COMPARE
Panel-Mount Standard Connector, Type-T, Round, Female, 1/Ea
₹1,700.00INR / Each
COMPARE
Panel-Mount Miniconnector, Type-J, Female, Rectangular, 1/Ea
₹990.00INR / Each
COMPARE
Panel-Mount Miniconnector, Type-K, Female, Rectangular, 1/Ea
₹990.00INR / Each
COMPARE
Panel-Mount Miniconnector, Type-T, Female, Rectangular, 1/Ea
₹990.00INR / Each
COMPARE
Panel-Mount Standard Connector, Type-J, Female, Rectangular, 1/Ea
₹1,400.00INR / Each
COMPARE
Panel-Mount Standard Connector, Type-K, Female, Rectangular, 1/Ea
₹1,400.00INR / Each
COMPARE
Panel-Mount Standard Connector, Type-T, Female, Rectangular, 1/Ea
₹1,400.00INR / Each
Showing 1- 12 of 12