Digi-Sense Panel-Mount Standard Connector, Type-J, Female, Rectangular, 1/Ea
Representative image only.
Digi-Sense Panel-Mount Standard Connector, Type-J, Female, Rectangular, 1/Ea
Digi-Sense | Item: 1852730
Round or rectangular shape to fit your instrument panel
₹1,800.00INR / Each
₹1,800.00INR / Each
Product Description
Key Features
  • Glass-filled nylon construction provides rigidity, strength, and surface hardness
  • Molded barriers prevent short circuit
  • 6 month warranty
More About this Item
Mount connectors to your panel using the two screws located on the front of the jack. Each connector is ANSI color-coded by thermocouple type: black (J), yellow (K), or blue (T). Choose between flat-bladed miniconnectors and round-pinned standard connectors.
6 Month Warranty
Specifications
  • Connector TypeJack
  • Special InformationGlass-filled nylon body
  • Max Temperature (° F)392
  • Max Temperature (° C)200
  • DescriptionPanel-Mount Standard Connector, Type-J, Female, Rectangular, 1/Ea