Digi-Sense High Temperature Mini Thermocouple Connectors
Handles extreme environments up to 650°F (343°C)!
  • Molded barriers prevent short circuit and the solid pin design ensures good contact even in demanding applications
  • Miniature connectors are suited for small wires and mineral insulated cables
  • Connector-body color is brown, calibration type (J or K) is printed on the body
High temperature connector-body color is brown, calibration indication printed on body.
1,800.00 - ₹2,400.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Miniature Type-K Thermocouple Male Connector, 2 Pin
₹1,800.00INR / Each
COMPARE
Miniature Type-J Thermocouple Male Connector, 2 Pin
₹1,800.00INR / Each
COMPARE
Miniature Type-K Thermocouple Female Connector, 2 Pin
₹1,800.00INR / Each
COMPARE
Miniature Type-J Thermocouple Female Connector, 2 Pin
₹1,800.00INR / Each
COMPARE
Standard Type-K Thermocouple Male Connector, 2 Pin, Hollow
₹2,400.00INR / Each
COMPARE
Standard Type-J Thermocouple Male Connector, 2 Pin, Hollow
₹2,400.00INR / Each
COMPARE
Standard Type-K Thermocouple Female Connector, 2 Pin
₹2,400.00INR / Each
COMPARE
Standard Type-J Thermocouple Female Connector, 2 Pin
₹2,400.00INR / Each
Showing 1- 8 of 8