Digi-Sense General-Purpose Thermocouple Connectors
Jacks and plugs good to 300°F (150°C)
  • Glass-filled nylon construction with molded barriers to prevent short circuit
  • Secure thermocouple wire into positive/negative terminals with screws
  • Each connector comes in ANSI color coded material: type J-black, type K-yellow, and type T-blue
Each connector comes in ANSI color coded material: type J-black, type K-yellow, and type T-blue.
1,600.00 - ₹7,200.00INR / Pkg of 5
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Thermocouple Connector, Mini, Type-T, Female Jack; 5/pk
₹4,900.00INR / Pkg of 5
COMPARE
Thermocouple Connector, Standard, Type-K, Male Plug; 1/ea
₹1,600.00INR / Each
COMPARE
Thermocouple Connector, Standard, Type-K, Male Plug; 5/pk
₹7,200.00INR / Pkg of 5
COMPARE
Thermocouple Connector, Standard, Type-J, Male Plug; 1/ea
₹1,600.00INR / Each
COMPARE
Thermocouple Connector, Standard, Type-J, Male Plug; 5/pk
₹7,200.00INR / Pkg of 5
COMPARE
Thermocouple Connector, Standard, Type-T, Male Plug; 1/ea
₹1,600.00INR / Each
COMPARE
Thermocouple Connector, Standard, Type-T, Male Plug; 5/pk
₹7,200.00INR / Pkg of 5
COMPARE
Thermocouple Connector, Standard, Type-K, Female Jack; 1/ea
₹1,600.00INR / Each
COMPARE
Thermocouple Connector, Standard, Type-K, Female Jack; 5/pk
₹7,200.00INR / Pkg of 5
COMPARE
Thermocouple Connector, Standard, Type-J, Female Jack; 1/ea
₹1,600.00INR / Each
COMPARE
Thermocouple Connector, Standard, Type-J, Female Jack; 5/pk
₹7,200.00INR / Pkg of 5
COMPARE
Thermocouple Connector, Standard, Type-T, Female Jack; 1/ea
₹2,100.00INR / Each
Showing 1- 12 of 12