Digi-Sense Thermocouple Connector, Standard, Type-K, Female Jack; 5/pk
Representative image only.
Digi-Sense Thermocouple Connector, Standard, Type-K, Female Jack; 5/pk
Digi-Sense | Item: 1852708
Jacks and plugs good to 300°F (150°C)
₹7,200.00INR / Pkg of 5
₹7,200.00INR / Pkg of 5
Buy It Together
Current Item
Digi-Sense Thermocouple Connector, Standard, Type-K, Female Jack; 5/pk
Digi-Sense Thermocouple Connector, Standard, Type-K, Female Jack; 5/pk
1852708
₹7,200.00INR / Pkg of 5
+
Traceable Monitoring Hygrometer
2025030
₹6,700.00INR / Each
=
Total Price 13900.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Glass-filled nylon construction with molded barriers to prevent short circuit
  • Secure thermocouple wire into positive/negative terminals with screws
  • Each connector comes in ANSI color coded material: type J-black, type K-yellow, and type T-blue
  • 6 month warranty
More About this Item
Each connector comes in ANSI color coded material: type J-black, type K-yellow, and type T-blue.
6 Month Warranty
Specifications
  • Connector TypeK
  • Special InformationGlass filled Nylon body
  • Max Temperature (° F)300
  • Max Temperature (° C)149
  • DescriptionThermocouple Connector, Standard, Type-K, Female Jack; 5/pk