Digi-Sense Economical Thermocouple Bolt-On Probes with Miniconnector
Economical Bolt-On probes designed to be an easy and cost effective way to mount on a surface
  • Available in #6 (M3.5) or #8 (M4) screw sizes and different lead lenghts
  • Response Time: 2-4 seconds or greater
  • Each probe comes with a color coded ANSI mini connector or stripped leads: type J-black, type K-yellow, type T-blue, and type E-purple.
  • Custom designs or changes to this product are available upon request
Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220°F (104°C); PVC handles are rated to 200°F (93°C); Stainless Steel handles are rated at 450°F (232°C); Mini Connectors are rated to 392°F (200°C).
5,100.00 - ₹7,600.00INR / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Probe Type
Availability Pricing
COMPARE
900 J
₹6,900.00INR / Each
COMPARE
500 T
₹6,900.00INR / Each
COMPARE
900 K
₹5,100.00INR / Each
COMPARE
900 K
₹7,200.00INR / Each
COMPARE
500 T
₹5,600.00INR / Each
COMPARE
900 K
₹7,200.00INR / Each
COMPARE
500 T
₹7,200.00INR / Each
COMPARE
900 K
₹5,900.00INR / Each
COMPARE
900 J
₹7,600.00INR / Each
COMPARE
500 T
₹5,900.00INR / Each
COMPARE
900 K
₹5,900.00INR / Each
COMPARE
900 J
₹7,600.00INR / Each
Showing 1- 12 of 12