Digi-Sense® Coiled Thermocouple Extension Cables with Mini-Connector
Thermocouple coiled extension cables for remote applications
  • Maximum temperature: 221° F (105° C)
  • Cables come in a variety of lengths
Each probe comes with color coded ANSI standard connectors: type J-black, type K-yellow, and type T-blue. Custom designs or changes to this product are available upon request.
6,600.00 - ₹7,400.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connector Type Description
Availability Pricing
COMPARE
J Coiled Extension Cable, Type J, Male to Female Mini-Connector, 5 ft Length
₹6,600.00INR / Each
COMPARE
K Coiled Extension Cable, Type K, Male to Female Mini-Connector, 5 ft Length
₹6,600.00INR / Each
COMPARE
T Coiled Extension Cable, Type T, Male to Female Mini-Connector, 5 ft Length
₹7,400.00INR / Each
COMPARE
J Coiled Extension Cable, Type J, Male to Male Mini-Connector, 5 ft Length
₹6,900.00INR / Each
COMPARE
K Coiled Extension Cable, Type K, Male to Male Mini-Connector, 5 ft Length
₹6,900.00INR / Each
COMPARE
T Coiled Extension Cable, Type T, Male to Male Mini-Connector, 5 ft Length
₹6,900.00INR / Each