Digi-Sense Blue Spirit Verification Bottle Standard Accuracy Thermometers - Celsius Scale
Enclosed bottle ensures accuracy and is ideal for use in environmental chambers
  • An individually serial number is printed on the thermometer stem
  • Nonmercury blue spirit liquid
Accurately monitor temperatures in freezers, refrigerators, incubators, ovens, and other environments where stability is required. All thermometers are supplied in a glass bead filled unbreakable plastic bottle and can be removed from the bottle for recalibration. Use the magnetic attachment with pressure-sensitive backing to attach the bottle to a metal wall or door.
Thermometers feature permanently fused markings, blue spirit liquid, white background glass; are also individually serial numbered. Include an statement of accuracy and a triangular-style antiroll fitting.
Call For Price
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Min Temperature (° C) Overall Length (mm)
Availability Pricing
COMPARE
25 -90 145
Call For Price
COMPARE
1 -30 135
Call For Price
COMPARE
20 -5 135
Call For Price
COMPARE
15 -5 125
Call For Price
COMPARE
45 10 125
Call For Price
COMPARE
50 15 135
Call For Price
COMPARE
130 20 145
Call For Price
COMPARE
200 50 145
Call For Price
Showing 1- 8 of 8