Digi-Sense Blue Spirit Certified Glass Thermometers, Fahrenheit-scale
NIST-Traceable individually calibrated thermometers feature precise and accurate temperature measurement
  • Nonmercury blue spirit liquid
  • NIST-certified detailed, laminated report of calibration included
These thermometers are ideal for use in applications where a very precise accurate temperature measurement is required. Thermometers are individually calibrated to NIST standards at specific temperatures and supplied with an NIST report of calibration stating corrections to one-tenth the smallest division.
Thermometers feature triangular anti-roll fitting, permanently fused markings, blue spirit liquid, white background glass; are also individually serial numbered. Include an NIST-traceable statement of accuracy and a triangular-style antiroll fitting.
46,100.00 - ₹55,200.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Min Temperature (° F) Overall Length (mm)
Availability Pricing
COMPARE
124 30 460
₹55,200.00INR / Each
COMPARE
300 0 300
₹46,100.00INR / Each