Dickson 3" Compact Temperature Recorders
High accuracy and resolution for regulatory compliance monitoring or quality control
Use these compact temperature recorders for monitoring and recording data in almost any location. These recorders are calibratable for accurate recording in refrigerators, freezers, incubators, and more. Units are °F/°C and 24-hour or 7-day selectable with a sturdy case that protects the recorders from damage in harsh environments. Units are wall-mountable or free-standing depending on your application requirements.
113,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Chart Diameter (in) Max Temperature (° F)
Availability Pricing
Dickson SC367 3" Temperature Chart Recorder; -14 to 32 F (-25 to 0 C) 8001400 Mfr # SC367
COMPARE
3 32
₹113,000.00INR / Each
Dickson SC377 3" Temperature Chart Recorder; 0 to 50°F (-15 to 10°C) 8001402 Mfr # SC377
COMPARE
3 50
₹113,000.00INR / Each
Dickson SC386 3" Temperature Chart Recorder; 22 to 68°F (-5 to 20°C) 8001404 Mfr # SC386
COMPARE
3 68
₹113,000.00INR / Each
Dickson SC387 3" Temperature Chart Recorder; 50 to 96°F (10 to 35°C) 8001406 Mfr # SC387
COMPARE
3 96
₹113,000.00INR / Each