Diba Omnifit® FKM O-Rings for Cap System Fittings and Valves
Created tight seal, holds tube in place
  • Constructed of durable FKM
Use where chemical resistance is less important and when the exact OD of the tube is not known. O-rings compress into the cone angle in the body of the fitting so when the cap is tightened a seal is created and the tube is held in place.
1,300.00INR / Pkg of 10
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Compatible With
Availability Pricing
Diba Omnifit® O-Ring for Connector and Valve Caps, small; 10/pk WW-21939-44 Mfr # 001401
COMPARE
Omnifit® Cap Style Fittings (1 to 2.5 mm tube OD)
₹1,300.00INR / Pkg of 10
Diba Omnifit® O-Ring for Connector and Valve Caps, large; 10/pk WW-21939-45 Mfr # 001402
COMPARE
Omnifit® Cap Style Fittings (1.5 to 4.0 mm tube OD)
₹1,300.00INR / Pkg of 10