Diba Omnifit® O-Ring for Connector and Valve Caps, large; 10/pk
Representative image only.
Diba Omnifit® O-Ring for Connector and Valve Caps, large; 10/pk
Diba | Mfr: 1402| Item: 2193945
Created tight seal, holds tube in place
₹2,000.00INR / Pkg of 10
₹2,000.00INR / Pkg of 10
Product Description
Key Features
  • Constructed of durable FKM
More About this Item
Use where chemical resistance is less important and when the exact OD of the tube is not known. O-rings compress into the cone angle in the body of the fitting so when the cap is tightened a seal is created and the tube is held in place.
Specifications
  • MaterialFKM
  • Compatible WithOmnifit® Cap Style Fittings (1.5 to 4.0 mm tube OD)
  • DescriptionOmnifit® O-Ring for Connector and Valve Caps, large; 10/pk