Diba Omni-Lok™ M6 ISO(M) and 5/16"-24 UNF(M) Compact Head Fittings for Type P/S Ferrules, PP
Permanently attached ferrules are ideal for repeated connect/disconnect
  • Free spinning fitting eliminates tubing from twisting
  • A recess in the fitting houses the ferrule allowing maximum thread engagement with the port
Type P ferrule has inner PTFE sealing face, the only material in the fluid path. Two-piece type S ferrule consists of an inner PEEK or ETFE sealing face surrounded by a stainless steel lock ring, with only the inner portion in the fluid path. Choose type P for higher pressures, type S for higher temperatures. Select glass-filled polypropylene compact head fittings available in a range of colors for easy identification.
2,800.00 - ₹4,800.00INR / Pkg of 10
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Connection Type Description
Availability Pricing
Diba Omni-Lok™ Compact Tubing End Fitting, Type S/P, 1/16"OD, M6 ISO(M), black PP; 10/pk WW-21942-16 Mfr # 008NF16-YC6B
COMPARE
1/16 Compression x ISO(M) Omni-Lok™ Compact Tubing End Fitting, Type S/P, 1/16\"OD, M6 ISO(M), black PP; 10/pk
₹2,800.00INR / Pkg of 10
Diba Omni-Lok™ Compact Tubing End Fitting, Type S/P, 1/8"OD, M6 ISO(M), black PP; 10/pk WW-21942-37 Mfr # 008NF32-YC6B
COMPARE
1/8 Compression x ISO(M) Omni-Lok™ Compact Tubing End Fitting, Type S/P, 1/8\"OD, M6 ISO(M), black PP; 10/pk
₹2,800.00INR / Pkg of 10
Diba Omni-Lok™ Compact Tubing End Fitting, Type S/P, 3/16"OD, 5/16"-24 UNF(M), black PP; 10/pk WW-21942-39 Mfr # 008NF47-YC7B
COMPARE
3/16 Compression x UNF(M) Omni-Lok™ Compact Tubing End Fitting, Type S/P, 3/16\"OD, 5/16\"-24 UNF(M), black PP; 10/pk
₹4,800.00INR / Pkg of 10