Diba Dibafit™ ETFE Inverted Cones (Ferrules)
Chemically inert ETFE cone is the only component in the fluid path
  • Constructed of durable ETFE
Utilize these ETFE inverted contes in your chromatography fluid path.
2,800.00 - ₹8,500.00INR / Pkg of 10
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Compression Size / Tube Od (Port 1) (mm) Description
Availability Pricing
Dibafit™ Inverted Cone (Ferrule), ETFE, 1/8" (3.2 mm); 10/pk WW-21939-59 Mfr # 001545
COMPARE
1/8 3.2 Inverted Cone (Ferrule), ETFE, 1/8\" (3.2 mm); 10/pk
₹2,800.00INR / Pkg of 10
Dibafit™ Short Inverted Cone (Ferrule), 1/16" OD tubing, ETFE; 10/pk WW-21940-43 Mfr # 008CZ16
COMPARE
1/16 1.57 Short Inverted Cone (Ferrule), 1/16\" OD tubing, ETFE; 10/pk
₹2,800.00INR / Pkg of 10
Dibafit™ Short Inverted Cone (Ferrule), 1.8 to 2.0 mm OD tubing, ETFE; 10/pk WW-21940-44 Mfr # 008CZ19
COMPARE
5/64 2 Short Inverted Cone (Ferrule), 1.8 to 2.0 mm OD tubing, ETFE; 10/pk
₹2,800.00INR / Pkg of 10
Dibafit™ Short Inverted Cone (Ferrule), 2.5 mm OD tubing, ETFE; 10/pk WW-21940-45 Mfr # 008CZ25
COMPARE
3/32 2.5 Short Inverted Cone (Ferrule), 2.5 mm OD tubing, ETFE; 10/pk
₹2,800.00INR / Pkg of 10
Dibafit™ Short Inverted Cone (Ferrule), 3.0 mm OD tubing, ETFE; 10/pk WW-21940-46 Mfr # 008CZ30
COMPARE
1/8 3 Short Inverted Cone (Ferrule), 3.0 mm OD tubing, ETFE; 10/pk
₹2,800.00INR / Pkg of 10
Dibafit™ Short Inverted Cone (Ferrule), 1/8" OD tubing, ETFE; 10/pk WW-21940-47 Mfr # 008CZ32
COMPARE
1/8 3.2 Short Inverted Cone (Ferrule), 1/8\" OD tubing, ETFE; 10/pk
₹2,800.00INR / Pkg of 10
Dibafit™ Short Inverted Cone (Ferrule), 3/16" OD tubing, ETFE; 10/pk WW-21940-48 Mfr # 008CZ47
COMPARE
3/16 4.8 Short Inverted Cone (Ferrule), 3/16\" OD tubing, ETFE; 10/pk
₹8,500.00INR / Pkg of 10
Showing 1- 7 of 7