Dibafit™ Short Inverted Cone (Ferrule), 3/16" OD tubing, ETFE; 10/pk
Representative image only.
Dibafit™ Short Inverted Cone (Ferrule), 3/16" OD tubing, ETFE; 10/pk
Diba | Mfr: 008CZ47| Item: 2194048
Chemically inert ETFE cone is the only component in the fluid path
₹12,600.00INR / Pkg of 10
₹12,600.00INR / Pkg of 10
Product Description
Key Features
  • Constructed of durable ETFE
More About this Item
Utilize these ETFE inverted contes in your chromatography fluid path.
Specifications
  • Fitting MaterialETFE
  • Compression Size / Tube Od (Port 1) (in)3/16
  • Compression Size / Tube Od (Port 1) (mm)4.8
  • Max Operating Pressure (PSI)200
  • DescriptionShort Inverted Cone (Ferrule), 3/16" OD tubing, ETFE; 10/pk