Diba Click-N-Seal® Fittings with M6 ISO(M) Thread for Dibafit™ Type P/S Ferrule System
Permanently attached, chemically inert ferrule is ideal for repeated connect/disconnect
  • Excellent for high-pressure application
  • When the cap clicks, you know the seal is tight
  • Self-torquing fittings create a secure seal without the use of any tools
Permanently attached ferrule allow fittings for use in high-pressure applications. As the fitting is tightened, the resistance of the cap increases until the correct torque is reached. The fitting can spin freely from tubing, preventing tubing from twisting. A recess in the fitting houses the ferrule, allowing maximum thread engagement with the port. Select fittings from acetal, polycarbonate (PC), or PEEK™ material of construction. PEEK fittings are compact in design and provide excellent chemical resistance.
Both Type P and Type S ferrules fits permanently onto the outside of semi-rigid tubing. Type P system uses a one-piece ferrule with only the inner PTFE sealing face in the fluid path. Type S system uses a two-piece ferrule made up of an inner PEEK or ETFE sealing surface surrounded by a stainless steel lock ring with only the inner portion in the fluid path.
3,800.00 - ₹12,200.00INR / Pkg of 6
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Compression Size / Tube Od (Port 1) (in) Fitting Material
Availability Pricing
Diba Click-N-Seal™ Plus Tubing End Fitting, Type S/P, 1/16" OD tubing, M6 ISO(M), white acetal; 10/pk WW-21941-93 Mfr # 008NF16-AT6H
COMPARE
White 1/16 Acetal
₹6,000.00INR / Pkg of 6
Diba Click-N-Seal™ Tubing End Fitting, Type S/P, 1/16" OD tubing, M6 ISO(M), black PC; 10/pk WW-21941-97 Mfr # 008NF16-CS6B
COMPARE
Black 1/16 Polycarbonate
₹3,800.00INR / Pkg of 10
Diba Click-N-Seal™ Tubing End Fitting, Type S/P, 1/16" OD tubing, M6 ISO(M), white PC; 10/pk WW-21941-98 Mfr # 008NF16-CS6H
COMPARE
White 1/16 Polycarbonate
₹3,800.00INR / Pkg of 10
Diba Click-N-Seal™ Tubing End Fitting, Type S/P, 1/16" OD tubing, M6 ISO(M), red PC; 10/pk WW-21941-99 Mfr # 008NF16-CS6R
COMPARE
Red 1/16 Polycarbonate
₹3,800.00INR / Pkg of 10
Diba Click-N-Seal™ Ultra Tubing End Fitting, Type S/P, 1/16" OD tubing, M6 ISO(M), natural PEEK™; 10/pk WW-21942-03 Mfr # 008NF16-KT6L
COMPARE
Natural 1/16 PEEK
₹12,200.00INR / Pkg of 10
Diba Click-N-Seal™ Plus Tubing End Fitting, Type S/P, 1/8" OD tubing, M6 ISO(M), acetal; 6/pk WW-21942-19 Mfr # 008NF32-AT6H
COMPARE
White 1/8 Acetal
₹6,000.00INR / Pkg of 6
Diba Click-N-Seal™ Tubing End Fitting, Type S/P, 1/8" OD tubing, M6 ISO(M), black PC; 10/pk WW-21942-23 Mfr # 008NF32-CS6B
COMPARE
Black 1/8 Polycarbonate
₹3,800.00INR / Pkg of 10
Diba Click-N-Seal™ Tubing End Fitting, Type S/P, 1/8" OD tubing, M6 ISO(M), white PC; 10/pk WW-21942-24 Mfr # 008NF32-CS6H
COMPARE
White 1/8 Polycarbonate
₹3,800.00INR / Pkg of 10
Diba Click-N-Seal™ Tubing End Fitting, Type S/P, 1/8" OD tubing, M6 ISO(M), red PC; 10/pk WW-21942-25 Mfr # 008NF32-CS6R
COMPARE
Red 1/8 Polycarbonate
₹3,800.00INR / Pkg of 10
Diba Click-N-Seal™ Ultra Tubing End Fitting, Type S/P, 1/8" OD tubing, M6 ISO(M), natural PEEK™; 10/pk WW-21942-27 Mfr # 008NF32-KT6L
COMPARE
Natural 1/8 PEEK
₹11,500.00INR / Pkg of 10
Showing 1- 10 of 10