Diba Click-N-Seal™ Tubing End Fitting, Type S/P, 1/8" OD tubing, M6 ISO(M), red PC; 10/pk
Representative image only.
Diba Click-N-Seal™ Tubing End Fitting, Type S/P, 1/8" OD tubing, M6 ISO(M), red PC; 10/pk
Diba | Mfr: 008NF32-CS6R| Item: 2194225
Permanently attached, chemically inert ferrule is ideal for repeated connect/disconnect
₹5,600.00INR / Pkg of 10
₹5,600.00INR / Pkg of 10
Buy It Together
Current Item
Diba Click-N-Seal™ Tubing End Fitting, Type S/P, 1/8" OD tubing, M6 ISO(M), red PC; 10/pk
Diba Click-N-Seal™ Tubing End Fitting, Type S/P, 1/8" OD tubing, M6 ISO(M), red PC; 10/pk
2194225
₹5,600.00INR / Pkg of 10
+ =
Total Price 40400.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Excellent for high-pressure application
  • When the cap clicks, you know the seal is tight
  • Self-torquing fittings create a secure seal without the use of any tools
More About this Item
Permanently attached ferrule allow fittings for use in high-pressure applications. As the fitting is tightened, the resistance of the cap increases until the correct torque is reached. The fitting can spin freely from tubing, preventing tubing from twisting. A recess in the fitting houses the ferrule, allowing maximum thread engagement with the port. Select fittings from acetal, polycarbonate (PC), or PEEK™ material of construction. PEEK fittings are compact in design and provide excellent chemical resistance.
Both Type P and Type S ferrules fits permanently onto the outside of semi-rigid tubing. Type P system uses a one-piece ferrule with only the inner PTFE sealing face in the fluid path. Type S system uses a two-piece ferrule made up of an inner PEEK or ETFE sealing surface surrounded by a stainless steel lock ring with only the inner portion in the fluid path.
Specifications
  • Fitting MaterialPolycarbonate
  • Connection TypeCompression x ISO(M)
  • Compression Size / Tube Od (Port 1) (in)1/8
  • Compression Size / Tube Od (Port 1) (mm)3.2
  • Thread Size ISO Metric (M) (Port 2) (mm)6
  • ColorRed
  • Max Operating Pressure (PSI)Type P with PTFE/SS ferrule: 1000 psiType P with PTFE/PEEK™ ferrule: 600 psiType S ferrule: 500 psi
  • DescriptionClick-N-Seal™ Tubing End Fitting, Type S/P, 1/8" OD tubing, M6 ISO(M), red PC; 10/pk