Diba Click-N-Seal® Adapters with Flat-Bottom 1/4"-28 UNF(M) Thread for Softwall Tubing
Hand tighten fittings without tools to create a secure seal
  • When the cap clicks, you know the seal is tight
These unique self-torquing fittings create a secure thread to barb connection without the use of any tools. As the fitting is tightened, the resistance of the cap increases until the correct torque is reached. Select from polycarbonate (PC) or PEEK™ material of construction. PEEK fittings are compact in design and provide excellent chemical resistance.
6,400.00 - ₹14,900.00INR / Pkg of 10
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Color Fitting Material
Availability Pricing
Diba Click-N-Seal™ Adapter, 1/4"-28 UNF(M) flat bottom to 1/16" ID, black PC; 10/pk WW-21941-20 Mfr # 008NB16-CS5B
COMPARE
1/16 Black Polycarbonate
₹6,400.00INR / Pkg of 10
Diba Click-N-Seal™ Adapter, 1/4"-28 UNF(M) flat bottom to 1/16" ID, white PC; 10/pk WW-21941-21 Mfr # 008NB16-CS5H
COMPARE
1/16 White Polycarbonate
₹6,400.00INR / Pkg of 10
Diba Click-N-Seal™ Adapter, 1/4"-28 UNF(M) flat bottom to 1/16" ID, red PC; 10/pk WW-21941-22 Mfr # 008NB16-CS5R
COMPARE
1/16 Red Polycarbonate
₹6,400.00INR / Pkg of 10
Diba Click-N-Seal™ Ultra Adapter, 1/4"-28 UNF(M) flat bottom to 1/16" ID, natural PEEK™; 10/pk WW-21941-26 Mfr # 008NB16-KT5L
COMPARE
1/16 Natural PEEK
₹14,900.00INR / Pkg of 10
Diba Click-N-Seal™ Adapter, 1/4"-28 UNF(M) flat bottom to 1/8" ID, black PC; 10/pk WW-21941-42 Mfr # 008NB32-CS5B
COMPARE
1/8 Black Polycarbonate
₹6,400.00INR / Pkg of 10
Diba Click-N-Seal™ Adapter, 1/4"-28 UNF(M) flat bottom to 1/8" ID, red PC; 10/pk WW-21941-44 Mfr # 008NB32-CS5R
COMPARE
1/8 Red Polycarbonate
₹6,400.00INR / Pkg of 10
Diba Click-N-Seal™ Ultra Adapter, 1/4"-28 UNF(M) flat bottom to 1/8" ID, natural PEEK™; 10/pk WW-21941-50 Mfr # 008NB32-KT5L
COMPARE
1/8 Natural PEEK
₹14,900.00INR / Pkg of 10
Diba Click-N-Seal™ Adapter, 1/4"-28 UNF(M) flat bottom to 5/32" ID, black PC; 10/pk WW-21941-53 Mfr # 008NB40-CS5B
COMPARE
5/32 Black Polycarbonate
₹6,400.00INR / Pkg of 10
Diba Click-N-Seal™ Adapter, 1/4"-28 UNF(M) flat bottom to 5/32" ID, white PC; 10/pk WW-21941-54 Mfr # 008NB40-CS5H
COMPARE
5/32 White Polycarbonate
₹6,400.00INR / Pkg of 10
Diba Click-N-Seal™ Adapter, 1/4"-28 UNF(M) flat bottom to 5/32" ID, red PC; 10/pk WW-21941-55 Mfr # 008NB40-CS5R
COMPARE
5/32 Red Polycarbonate
₹6,400.00INR / Pkg of 10
Showing 1- 10 of 10