Diba Click-N-Seal™ Adapter, 1/4"-28 UNF(M) flat bottom to 1/8" ID, black PC; 10/pk
Representative image only.
Diba Click-N-Seal™ Adapter, 1/4"-28 UNF(M) flat bottom to 1/8" ID, black PC; 10/pk
Diba | Mfr: 008NB32-CS5B| Item: 2194142
Hand tighten fittings without tools to create a secure seal
₹9,500.00INR / Pkg of 10
₹9,500.00INR / Pkg of 10
Product Description
Key Features
  • When the cap clicks, you know the seal is tight
More About this Item
These unique self-torquing fittings create a secure thread to barb connection without the use of any tools. As the fitting is tightened, the resistance of the cap increases until the correct torque is reached. Select from polycarbonate (PC) or PEEK™ material of construction. PEEK fittings are compact in design and provide excellent chemical resistance.
Specifications
  • Fitting MaterialPolycarbonate
  • Connection TypeBarb x UNF(M)
  • Thread size UNF(M) (port 2) (InchShort)1/4-28
  • Barb Size / Tubing Id (Port 1) (mm)3.2
  • Barb Size / Tubing Id (Port 1) (in)1/8
  • ColorBlack
  • DescriptionClick-N-Seal™ Adapter, 1/4"-28 UNF(M) flat bottom to 1/8" ID, black PC; 10/pk