Depth Gauges

Precision depth measurement to 22"
Use these gauges for measuring the depth of small holes and fabricated articles with grooved or elevated surfaces. Reliable, high accuracy depth measurements in micrometer or gauge style.
Call For Price

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max range (in)
Availability Pricing
Fowler 52-225-226 Digital Counter Depth Micrometer; 0 to 6" WW-97151-10 Mfr # 52-225-226
COMPARE
6
Call For Price
Fowler 54-225-456 Electronic Depth Gauge/Micrometer; 0 to 6" WW-97151-20 Mfr # 54-225-456
COMPARE
6
Call For Price
Fowler 52-125-006-1 Dial/Analog Depth Gauge; 0 to 22" WW-97151-30 Mfr # 52-125-006-1
COMPARE
22
Call For Price
Fowler 54-125-777 Electronic Depth Gauge; 0 to 22" WW-97151-40 Mfr # 54-125-777
COMPARE
22
Call For Price
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat