Deep-Ledge Utility Carts
Shelves feature a deep, 2 3/4” lip to contain spills
These carts are fabricated from a tough, high-density polyethylene and provide an excellent chemical resistance to most substances, including most acids and bases. The two- and three-shelf carts have been manufactured with Microban®, an antimicrobial product that protects shelves from bacteria, mold, and mildew.
Shelves are smooth and easily wipe clean. The middle shelves of the three- and four-shelf model adjusts at 1" increments. All models come with 5" -dia swivel casters.
66,200.00 - ₹75,700.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (lbs) Number of Shelves
Availability Pricing
Metro Carts BC2030-2DMB Deep-Ledge Utility Cart, 2 Shelf, Blue, 21.5x41x32.75", 400 lbs Cap 4730016 Mfr # BC2030-2DMB
COMPARE
400 2
₹66,200.00INR / Each
Metro Carts BC2030-3DMB Deep-Ledge Utility Cart, 3 Shelf, 21.5x41x32.75", 400 lbs Cap 4730018 Mfr # BC2030-3DMB
COMPARE
400 3
₹75,700.00INR / Each