Cryo-Babies and Cryo-Tags Labels
Cryo-Babies cryogenic labels are constructed of flexible polyvinyl and feature chemical-resistantce. Labels withstand temperatures down to -196°C and adhere to plastics, glass, and metal. Mark using any writing instrument. Suitable for autoclaving, boiling, water baths, or dry heat up to 150°C without peeling, cracking, or degrading. Lables can survive soaking at room temperature or 100% relative humidity at 35°C for at least 24 hours.
18,500.00INR / Roll of 1000
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Width (in)
Availability Pricing
COMPARE
1.2812 0.2812
₹18,500.00INR / Roll of 1000
COMPARE
1.2812 0.75
₹18,500.00INR / Roll of 1000